Learn More
Sigma Aldrich Fine Chemicals Biosciences Anti-NeuN Antibody, clone A60, 500 μG
Supplier:  Sigma Aldrich Fine Chemicals Biosciences MAB377
Encompass
Anti-NeuN Antibody, clone A60 detects level of NeuN and has been published and validated for use in FC, IC, IF, IH, IH(P), IP and WB. Synonym: Neuron-Specific Nuclear Protein, Neuna60, A60.
Catalog No. NC1721567