Learn More
STEMCELL Technologies Anti-Oligodendrocyte Marker O4 Antibody, Clone 81, Size: 50 µg
SDP
Supplier:  STEMCELL Technologies 60053
Encompass
Clone 81 is a mouse monoclonal antibody that reacts with the O4 antigen, a sulfated galactocerebroside (sulfatide). Verified for use with immunocytochemistry assays, use this antibody in applications requiring the detection of human, mouse, and rat oligodendrocyte marker O4. O4 is formed during postnatal development and is a marker for cell bodies and processes of oligodendrocytes types I and II. O4 is expressed in pro-oligodendrocytes, however, it is not found on O-2A-progenitor cells. Abnormal expression of O4 has been associated with various disorders, including neurofibromatosis and schizophrenia. Visit STEMCELL.com for more information and related products.
Catalog No. 50-197-5314
Only null left
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov