Learn More
Vector Laboratories ANTI P23 /3D1/ ANTIBDY 5ML
SDP
Supplier:  Vector Laboratories EAZ013
Encompass
These mouse monoclonal antibodies MAbs were produced against affinity-purified Cryptosporidium parvum /Iowa/ native antigens using sporozoite apical and surface pellicle-reactive MAbs C4A1 and C6B6 as binding ligands and recognize Cryptosporidium spp. oocysts or apical and surface pellicle antigens GP25-200 or P23.
Catalog No. NC2874128