Learn More
Vector Laboratories ANTI-P23 5B9 ANTIBDY 5ML
SDP
Supplier:  Vector Laboratories EAZ014
Encompass
These mouse monoclonal antibodies MAbs were produced against affinity-purified Cryptosporidium parvum /Iowa/ native antigens using sporozoite apical and surface pellicle-reactive MAbs C4A1 and C6B6 as binding ligands and recognize Cryptosporidium spp. oocysts or apical and surface pellicle antigens GP25-200 or P23.
Catalog No. NC2874131