Learn More
Sino Biological Anti-PARD6A Antibody, Rabbit Polyclonal 100 µL
Supplier:  Sino Biological 200175T32100
Encompass_Preferred
Produced in rabbits immunized with E. coli-derived Human PARD6A fragment, and purified by antigen affinity chromatography.
Catalog No. 50-007-48857