Learn More
ANP Technologies Inc ANTI-PEG MONOCLONAL IGM AB
SDP
Supplier:  ANP Technologies Inc 901010100UG
Encompass
Anti-PEG Monoclonal IgM Antibody (ANPEG-1).Description: Mouse monoclonal anti-PEG antibody/PEC antibody specifically binds to the repeating unit of PEG polymer
Catalog No. NC2241453