Learn More
Santa Cruz Biotechnology ANTI-PKR ANTIBODY (B-10)
Supplier:  Santa Cruz Biotechnology SC6282
Encompass
Anti-PKR Antibody (B-10) is a mouse monoclonal IgG2a κ PKR antibody provided at 200 µg/ml.
Catalog No. NC1996839