Learn More
Rockland Immunochemicals Anti-Potassium Chloride Cotransporter (KCC2) pS940 (Rabbit) Antibody - 612-401-E15
SDP
Supplier:  Rockland Immunochemicals 612401E15
Encompass_Preferred
Potassium Chloride Cotransporter pS940 Antibody detects Potassium Chloride Cotransporter pS940 protein. KCC2 is widely thought to be expressed exclusively in neurons where it is responsible for maintaining low intracellular chloride concentration to drive hyperpolarizing post-synaptic responses to the inhibitory neurotransmitters GABA and glycine. Serine 940 on KCC2 has been shown to be phosphorylated by PKC and has further been demonstrated to be the major site for PKC-dependent phosphorylation for full length KCC2 molecules when expressed in HEK-293 cells as phosphorylation of Ser940 increased the cell surface stability of KCC2 in this system by decreasing it’s rate of internalization from the plasma membrane. Anti-Potassium Chloride Cotransporter (KCC2) pS940 Antibody is ideal for investigators involved in Neuroscience and Cell Signaling research.
Catalog No. 50-105-8641