Learn More
Advanced ImmunoChemical, Inc. Monoclonal Mouse Anti-Human Procalcitonin (PCT), 0.2mg
Supplier:  Advanced ImmunoChemical, Inc. 2PCAL0.2MG
Encompass_Preferred
This item is manufactured by HyTest, Ltd and distributed by Advanced ImmunoChemical, Inc. Monoclonal mouse anti-human procalcitonin (PCT). MAbs:6F10, 27A3, 38F11, 44D9, 42, 14C12, 22A11, 18B7Hybridoma clones have been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with fragments of human PCT conjugated with carrier protein.Specificity:Human PCTMAb Isotypes:IgG1 for MAbs 6F10, 38F11, 14C12, 22A11, 18B7IgG2a for MAbs 27A3, 44D9 and 42Applications:Detection of human PCT in ELISA and Western blotting.Recommended pairs for sandwich immunoassay (capture-detection):16B5 – 42, 27A3 – 14A2, 14C12 – 14A2, 18B7 – 44D9, 38F11 – 22A11 (Note: MAbs 16B5 and 14A2 are under Cat.# 2-CALC)Purification:Chromatography on protein A SepharosePresentation:PBS, pH 7.4, 0.09 % sodium azide (NaN3)Storage Conditions:+ 4 °COther information:More antibodies against calcitonin under Cat.# 2-CALC. This product is sold as an antibody preparation for research use only.
Catalog No. 50-153-9865