Learn More
Advanced ImmunoChemical, Inc. ANTI-PSA MAB
Supplier:  Advanced ImmunoChemical, Inc. 2PSA1 MG
Encompass_Preferred
MONOCLONAL ANTIBODIES/MAbs in vitro:8A6ccMAbs in vivo:5A6, PS2, 1H12Hybridoma clones have been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with human PSA antigen purified from seminal plasma.MAb Isotypes:IgG1 for MAbs PS2, IgG2a for MAbs 5A6, 8A6cc, 1H12Applications:Detection of PSA antigen. Recommended pairs for detection of total PSA (capture-detection):Recommended pairs for detection of free PSA (capture-detection):5A6-8A6cc, Best pair: 8A6cc – 5A6MAbs can also be used in lateral flow.Purification:MAbs PS2 by ion exchange chromatography and other MAbs by chromatography on protein A SepharosePresentation:MAbs 5A6, 8A6, and 1H12 in PBS, pH 7.4, 0.1 % sodium azide (NaN3) MAbs PS2 in 0.15 M NaCl, 10 mM Tris, pH 7.5, 0.05% sodium azide (NaN3)Storage Conditions:+ 4 °C
Catalog No. NC3566226