Learn More
ECM Bioscience PTP1B (C-terminal region) Mouse Monoclonal Antibody, ECM Biosciences
Supplier:  ECM Bioscience PM2341
Encompass_Preferred
Clone M234 was generated from a recombinant protein corresponding to amino acids in the C-terminal region of human PTP1B. Accession #P18031. This antibody detects a 50 kDa* band corresponding to PTP1B in western blots of human A431, Jurkat, and HeLa cells. It does not detect PTP1B in rat PC12 or mouse SYF cells. Applications: WB, E, ICC; Species Reactivity: Hu; Size = 100 μl; Buffer = Mouse monoclonal antibody purified with protein A chromatography is supplied in 100µl phosphate-buffered saline, 50% glycerol, 1 mg/ml BSA, and 0.05% sodium azide. Store at –20°C. Stable for 1 year. Other Names = PTPN1; TCPTP/PTPN2
Catalog No. 50-206-8794