Learn More
Novus Biologicals Anti-Mouse Detection Module, 24 x 25 Caps
SDP
Supplier:  Novus Biologicals SWDM022
Encompass
Includes 3 Detection Modules. Each Detection Module includes Luminol-S, Peroxide, Antibody Diluent 2, Anti-Mouse Secondary Antibody, Streptavidin-HRP. Sufficient for 600 samples. Store the unopened product at 2 - 8 °C. Do not use past expiration date.
Catalog No. NC3591811