Learn More
Medchemexpress LLC Anti-Rabbit IgG for 50uL | 50uL
SDP
Supplier:  Medchemexpress LLC HYP8749650UL
Encompass_Preferred
Anti-Rabbit IgG for IP (HRP) is a conformation-specific secondary antibody that binds exclusively to native non-denatured IgG and does not recognize denatured IgG Conformation-specific secondary antibodies target spatial epitopes When antibodies are denatured by high temperatures they lose their native conformation and are no longer recognized by such antibodies neither the heavy chain nor the light chain is detected This effectively avoids interference from the heavy chain (55 kDa) and light chain (25 kDa) in immunoprecipitation (IP) experiments
Catalog No. 50-003-24481