Learn More
Rockland Immunochemicals Anti-RABBIT IgG (H&L) (GOAT) Antibody Peroxidase Conjugated (Min X Human Serum Proteins) - 611-1322-0500
SDP
Supplier:  Rockland Immunochemicals 61113220500
Encompass_Preferred
Anti-Rabbit IgG (H&L) generated in goat detects rabbit Immunoglobulin G. Both the Heavy and Light chains of the antibody molecule are present. Representing approximately 75% of serum immunoglobulins, IgG is the most abundant antibody isotype found in the circulation. IgG molecules are synthesized and secreted by plasma B cells. Secondary Antibodies are available in a variety of formats and conjugate types. When choosing a secondary antibody product, consideration must be given to species and immunoglobulin specificity, conjugate type, fragment and chain specificity, level of cross-reactivity, and host-species source and fragment composition. This Anti-Rabbit IgG is conjugated with Peroxidase.
Catalog No. 50-105-8537