Learn More
AbD Serotec ANTI RAT ED1
Supplier:  AbD Serotec MCA341R
Encompass
Mouse Anti Rat, Specificity: CD68, Product Type: Monoclonal Antibody, Clone: ED1, Isotype: IgG1, Quantity/Volume: 0.25mg, Purified IgG prepared by affinity chromatography on Protein A from tissue culture supernatant
Catalog No. NC9197930