Learn More
Rockland Immunochemicals Rat IgG (H&L) Antibody CY5 Conjugated Pre-Adsorbed 1 mg
SDP
Supplier:  Rockland Immunochemicals 612110120
Encompass
Anti-Rat IgG (H&L) CY5 Antibody generated in goat detects reactivity to Rat IgG. Secondary Antibodies are available in a variety of formats and conjugate types. When choosing a secondary antibody product consideration must be given to species and immunoglobulin specificity conjugate type fragment and chain specificity level of cross-reactivity and host-species source and fragment composition. This Anti-Rat IgG is conjugated to CY5.
Catalog No. 50-194-3931