Learn More
ECM Bioscience Anti-Rat IgG:HRP Donkey Polyclonal, ECM Biosciences
Supplier:  ECM Bioscience RS3101
Encompass_Preferred
This antibody has been pre-adsorbed with various immunoglobulins from non-rat species before affinity-purification using rat IgG coupled to agarose beads. Purified donkey polyclonal antibody was conjugated to horseradish peroxidase (HRP). This secondary reagent can be used to detect rat immunoglobulins in various antibody applications that utilize HRP readouts, such as western blot, ELISA, immunocytochemistry, and immunohistochemistry. The immunogen was The HRP-conjugated donkey polyclonal secondary reagent reacts with the heavy and light chains of rat IgG, and the light chains of other rat immunoglobulins. The secondary reagent has minimal cross-reactivity with bovine, chicken, goat, horse, human, mouse, rabbit, and sheep immunoglobulins. Accession #. Applications = WB, E, ICC, IHC; Species Reactivity = Rt; Size = 100 μl; Buffer = Donkey polyclonal antibody is supplied in 100µl PBS, 50% glycerol, and 1 mg/ml BSA.
Catalog No. 50-206-8825