Learn More
ECM Bioscience RCAN1/Dscr1 (a.a. 132-140) Rabbit Polyclonal Antibody, ECM Biosciences
Supplier:  ECM Bioscience RP3961
Encompass_Preferred
The antibody detects several isoforms of RCAN1 on SDS-PAGE immunoblots of mouse C2C12, rat PC12, and human Jurkat cells. The predominate isoform detected is the 38 kDa RCAN1L, but the antibody may also detect RCAN2 and RCAN3. The immunogen was RCAN1 synthetic peptide (coupled to carrier protein) corresponds to amino acids 132 to 140 in human RCAN1S (isoform 2). This sequence is highly conserved in rat and mouse RCAN1, and this sequence is found in all four isoforms of RCAN1. In addition, this sequence has significant homology to RCAN2 and RCAN3. Accession #P53805. Applications = WB, E, ICC; Species Reactivity = Hu, Rt, Ms; Size = 100 μl; Buffer = PBS, 50% glycerol, 1 mg/ml BSA, and 0.05% sodium azide. Store at –20°C. Stable for 1 year. Other Names = Dscr1, MCIP, RCAN1, calcipressin, Adapt78
Catalog No. 50-206-8824