Learn More
Sino Biological Anti-S100A9 Antibody, Mouse Monoclonal 100 µL
Supplier:  Sino Biological 11145MM20100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human S100A9 / CAGB / P14 (rh S100A9 / CAGB / P14; Catalog #11145-HNAE; Met1-Pro114; NP_002956.1). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-007-51139