Learn More
Sino Biological Anti-Serpin A1/Alpha-1-antitrypsin Antibody, Rabbit Polyclonal 100 µL
Supplier:  Sino Biological 10306T54100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human Serpin A1/Alpha-1-antitrypsin (rh Serpin A1/Alpha-1-antitrypsin; Catalog#10306-H08H; NP_000286.3; Met1-Lys418). Serpin A1/Alpha-1-antitrypsin specific IgG was purified by Human Serpin A1/Alpha-1-antitrypsin affinity chromatography.
Catalog No. 50-007-50135