Learn More
Sino Biological Anti-SerpinA3 Antibody, Rabbit Polyclonal 100 µL
Supplier:  Sino Biological 10307T48100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human SerpinA3 (rh SerpinA3; Catalog#10307-H08H; NP_001076.2; Met1-Ala423). SerpinA3 specific IgG was purified by Human SerpinA3 affinity chromatography.
Catalog No. 50-007-49480