Learn More
Immunology Consultants anti-SFTS Virus Antibody (Mouse - Clone #3G9) - Monoclonal
SDP
Supplier:  Immunology Consultants MSFTS55A3G9
Encompass_Preferred
This monoclonal antibody targets a distinct SFTS viral epitope, providing complementary detection to other clones. Clone 3G9 delivers strong affinity and low background in ELISA, Western blotting, and antigen‑capture workflows. MSFTS‑55A‑3G9 is ideal for diagnostic assay development, viral monitoring, and research on SFTS infection dynamics. Its consistent performance ensures reliable detection across diverse sample matrices.
Catalog No. 50-004-35401