Learn More
ECM Bioscience ShcA Rabbit Polyclonal Antibody, ECM Biosciences
Supplier:  ECM Bioscience SP1331
Encompass_Preferred
The antibody detects 46/52/66 kDa* bands corresponding to the molecular weight of ShcA variants on SDS-PAGE immunoblots of A431 cells. It also detects ShcA in human Jurkat, HeLa, and SKN-SH cells, as well as rat brain tissue and rabbit spleen fibroblasts. The immunogen was A recombinant protein that included amino acid residues within the C-terminal region of human ShcA. This sequence has high homology with similar regions in rat and mouse ShcA. Accession #P29353. Applications = WB, E, ICC; Species Reactivity = Hu, Rt, Ms, Rb; Size = 100 μl; Buffer = PBS, 50% glycerol, 1 mg/ml BSA, and 0.05% sodium azide. Store at –20°C. Stable for 1 year. Other Names = SH2 C1, SHC
Catalog No. 50-206-8857