Learn More
New England Biolabs, Inc. Anti-SNAP-tag Antibody (Polyclonal) – 100 µl
SDP
Supplier:  New England Biolabs, Inc. P9310S
Encompass
The Anti-SNAP-tag Antibody (Polyclonal) can be used for the detection of SNAP-tagged fusion proteins in Western blots.
Catalog No. 50-445-1