Learn More
Selleck Chemical LLC Anti-Sphingosine-1-phosphate
SDP
Supplier:  Selleck Chemical LLC A28571MG5
Encompass_Preferred
Anti-Sphingosine-1-phosphate is a human IgG1 monoclonal anti-Sphingosine-1-phosphate antibody that targets lysosphingolipids thereby lowering the concentration of S1P in the extracellular fluid MW 145 74 KD
Catalog No. 50-313-5259