Learn More
FUJIFILM BIOSCIENCES INC Anti SQSTM1/A170/p62 Rabbit 10
Supplier:  FUJIFILM BIOSCIENCES INC 01822141
Encompass_Preferred
Sequestosome 1 SQSTM1 /A170 mouse /p62 human /ZIP rat a ubiquitin-binding protein expresses oxidative stress-dependently Abnormality of SQSTM1 leads to bone metabolic disorder obesity and Type II diabetes SQSTM1 was reported to bind LC3 which regulates autophagosome formation The protein has attracted the attention of researchers because it is believed to induce a protein from ubiquitin-proteosome system UPS to the lysosome-dependent macroautophagy autophagy system which are two major intracellular pathways for protein degradation This antibody is useful for immunofluorescence immunohistochemistry and Western blotting
Catalog No. 50-004-50113