Learn More
ABclonal Technology SSTR2 Polyclonal Antibody,100UL,Validated in: WB,IHC
SDP
Supplier:  ABclonal Technology A3135
Encompass_Preferred
Immunogen: Recombinant fusion protein containing a sequence corresponding to amino acids 302-369 of human Somatostatin Receptor 2 (SSTR2) (NP_001041.1).•Host Species: Rabbit•Isotype: IgG•Species Reactivity:Human,Mouse,Rat•Validated Applications:WB,IHC•Swiss Prot: P30874•Gene ID:6752•Purification Method:Affinity purification•Storage: Store at -20℃. Avoid freeze / thaw cycles.|Buffer: PBS with 0.02% sodium azide,50% glycerol,pH7.3.This product is available in 20UL, 50UL, 100UL, and 200UL. Please contact customer service for pricing and availability on alternative sizes.
Catalog No. 50-164-8559