Learn More
Uline Uline Foam Roll | Pink | 1 EA
Supplier:  Uline S789
Encompass
Lightweight nonabrasive polyethylene foam provides excellent protection from static discharge in shipping, Great for circuit boards resistors and other electronic components, Cross perforated every 12, MEETS CID A-A-59135-CLASS 1 GRADE B
Catalog No. NC2261983