Learn More
Abcam Anti-Tau (MTBR-tau243) antibody [EPR29541-59]
Supplier:  Abcam AB3225541ML
Encompass_Preferred
Rabbit Recombinant Monoclonal TAU antibody. Suitable for WB, I-ELISA, IHC-P and reacts with Transfected cell lysate - Human, Human, Mouse, Recombinant fragment - Human, Synthetic peptide - Human, Recombinant fragment - Mouse samples.
Catalog No. NC3833701
Only null left
Add to Cart
Request a Quote