Learn More
TARGETMOL CHEMICALS INC ANTI-TB AGENT 1 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T103295MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. anti-TB agent 1 is a potent and orally active anti-tuberculosis agent (MICs < 2 nM against the Mtb strains H37Rv rRMP and rINH). purity: 98%
Catalog No. 50-258-1736