Learn More
KRISHGEN BIOSYSTEMS Immunoassay for the quantitative Determination of Anti-Ustekinumab antibodies in human serum and plasma. Assay Range: 10 - 640 ng/ml; Sensitivity: 5ng/ml.
Supplier:  KRISHGEN BIOSYSTEMS KBI2014
Encompass_Preferred
Pack Size: 1 x 96 wells. This ELISA is optimised to detect antibodies to ustekinumab in human samples and may be used for immunogenicity monitoring. If you would like to use animal samples, please contact us. Introduction: Ustekinumab is a human monoclonal antibody. It is directed against interleukin 12and interleukin 23, naturally occurring proteins that regulate the immune system and immune-mediated inflammatory disorders. Ustekinumab is approved in Canada, Europe and the United States to treat moderate to severe plaque psoriasis. ADA has been observed in preclinical and clinical studies, resulting in significant changes in toxicology, pharmacokinetics and efficacy. These effects result from the generation of drug-induced (neutralizing) autoantibodies against Etanercept and can be responsible for allergic reaction, or even anaphylactic shock. ELISA Features include a pre-coated plate with break-apart wells, standardisation and Precision CV <10%.
Catalog No. NC2747149