Learn More
Medchemexpress LLC Antibacterial agent 125 | 1274611-43-6 | C15H11ClN2O | 50 MG
SDP
Supplier:  Medchemexpress LLC HY15191850MG
Encompass_Preferred
Antibacterial agent 125 is an antibacterial agent with potent antimicrobial activity against clinically relevant Gram-positive pathogens, showing MIC50 values ranging from 0.25 - 8 μM. It can be utilized for research into antimicrobial resistance.
  • Demonstrates inhibitory activity against methicillin-resistant Staphylococcus aureus strains without inducing cytotoxicity in human cells.
  • Possesses high metabolic stability and an excellent safety profile.
  • Does not induce resistance after ten passages.
  • Has a moderate ability to inhibit biofilm formation.
Catalog No. 50-003-73903