Learn More
TOKU-E Erythromycin ReadyMade™ Solution, 114-07-8, 10 ML
Supplier:  TOKU-E E09610ML
Encompass_Preferred
Antibiotic, ReadyMade™, sterile-filtered, 1 mg/ml in water. Useful in cancer research, microbiology, cell culture, plant biology.
Catalog No. 50-255-1803