Learn More
TOKU-E Amphotericin B ReadyMade™ Solution, 397-89-3, 20 ML
Supplier:  TOKU-E A23420ML
Encompass_Preferred
Antibiotic, ReadyMade™, sterile-filtered, 0.25 mg/ml in water. Useful in microbiology, cell culture, plant biology.
Catalog No. 50-255-1836