Learn More
TOKU-E Tylosin ReadyMade™ Solution, 1401-69-0, 20 ML
Supplier:  TOKU-E T15020ML
Encompass_Preferred
Antibiotic, ReadyMade™, sterile-filtered, 8 mg/ml in 0.9% NaCl. Useful in cancer research, cell culture, plant biology.
Catalog No. 50-255-1750