Learn More
TOKU-E Kanamycin ReadyMade™ Solution, CAS, 10x100 ML
Supplier:  TOKU-E K03610X100ML
Encompass_Preferred
Antibiotic, ReadyMade™, sterile-filtered, 10 mg/ml in 0.9% NaCl. Useful in cancer research, cell culture, microbiology.
Catalog No. 50-255-1780