Learn More
Cayman Chemical ANTIBODY 1-A-N 1mg
SDP
Supplier:  Cayman Chemical 405001 MG
Encompass_Preferred
An ionizable cationic lipid; has been used in the generation of LNPs for siRNA delivery in vivo; LNPs containing 1-A-N and encapsulating siRNA targeting the gene encoding CD45 reduce its gene expression in splenic T cells in mice
Catalog No. 50-297-8403