Learn More
Cayman Chemical ANTIBODY 1-EBIO 100mg
SDP
Supplier:  Cayman Chemical 14575100 MG
Encompass_Preferred
An activator of Ca2+-sensitive K+ channels (EC50 = 490 µM in T84 human carcinoma cells); also activates the human intermediate conductance Ca2+-activated K+ channel (EC50 = 74 µM)
Catalog No. 50-297-2024