Learn More
Cayman Chemical ANTIBODY 11 R-HEDE 25ug
SDP
Supplier:  Cayman Chemical 3750525 UG
Encompass_Preferred
A mono hydroxy fatty acid derived from of 11Z,14Z-eicosadienoic acid in an LO-type reaction catalyzed by COX; spectrophotometric measurement of the conjugated diene is occasionally used to quantify COX activity
Catalog No. 50-291-5685