Learn More
Cayman Chemical ANTIBODY 11 S-HEPE 50ug
SDP
Supplier:  Cayman Chemical 3251050 UG
Encompass_Preferred
11(S)-HEPE is a monohydroxy fatty acid derived from EPA, theoretically by the action of COX. Although its arachidonate derived congener 11(S)-HETE has been isolated from cultured neutrophils, it is not known if similar incubations with EPA can produce 11(S)-HEPE.{1088}
Catalog No. 50-294-7085