Learn More
Cayman Chemical ANTIBODY 13-OxoDE 25ug
SDP
Supplier:  Cayman Chemical 3862025 UG
Encompass_Preferred
13-oxoODE is produced from 13-HODE by a NAD+-dependent dehydrogenase present in rat colonic mucosa.{184} 13-OxoODE stimulates cell proliferation when instilled intrarectally in rats.{2210} 13-OxoODE has also been detected in preparations of rabbit reticulocyte plasma and mitochondrial membranes, mostly esterified to phospholipids. Production of 13-oxoODE is putatively linked to the maturation of reticulocytes to erythrocytes through the activity of 15-LO.{2261,2262}
Catalog No. 50-291-8792