Learn More
Cayman Chemical ANTIBODY 15 S-HEPE 250ug
SDP
Supplier:  Cayman Chemical 32710250 UG
Encompass_Preferred
A monohydroxy fatty acid derived from EPA; serum levels are elevated in patients with asthma
Catalog No. 50-294-6538