Learn More
Cayman Chemical ANTIBODY 15 S-HpEPE 250ug
SDP
Supplier:  Cayman Chemical 42710250 UG
Encompass_Preferred
An active metabolite of EPA; inhibits IL-1β-induced increases in the levels of COX-2 in HPMECs at 0.01, 0.3, or 1 µM
Catalog No. 50-294-2751