Learn More
ACROBiosystems Monoclonal Anti-DNP antibody, Human IgG1 Isotype Control
Supplier: ACROBiosystems DNPM21MG
Antibody;1MG;Human IgG1/kappa;Mouse monoclonal antibody is produced from a hybridoma resulting from fusion of SP2/0 myeloma and B-lymphocytes obtained from a mouse immunized with DNP F(ab')2.; Dinitrophenyl (DNP) is a hapten that is not normally expressed in the target tissue. Therefore, anti-DNP antibody has no relevant specificity to a target antigen, and can be used as isotype control antibody to differentiate non-specific background signal from specific antibody signal in various in vitro and in vivo studies.Monoclonal Anti-DNP (Anti-Hapten) antibody, Human IgG1 was purified from HEK293 cell culture at ACRObiosystems, and specifically reacts with DNP (Dinitrophenyl) and DNP conjugated proteins.