Learn More
Cayman Chemical ANTIBODY 2 5-DmA 50mg
SDP
Supplier:  Cayman Chemical 1513150 MG
Encompass_Preferred
A natural triterpene which stimulates wound healing induces cell cycle arrest and apoptosis in breast cancer cells (IC50 5.9 UM for MCF-7 cells) blocks angiogenesis in glioblastomas down regulates BACE1 while increasing ADAM10 maturation in primary rat cortical neurons protects against neuronal damage and cognitive defects resulting from glutamate administration in vivo in mice
Catalog No. 50-297-0956