Learn More
Cayman Chemical ANTIBODY 2-D08 10mg
SDP
Supplier:  Cayman Chemical 2045910 MG
Encompass_Preferred
A synthetic flavone inhibitor of sumoylation; inhibits sumoylation completely in microfluidic electrophoretic mobility shift and kinetic assays at a concentration of 30 µM; inhibits sumoylation in breast cancer cell lines; disrupts the transfer of SUMO to the substrate; neuroprotective against Aβ aggregation and toxicity; has antioxidant properties
Catalog No. 50-298-1679