Learn More
Cayman Chemical ANTIBODY 2-thIo PAF 5mg
SDP
Supplier:  Cayman Chemical 609455 MG
Encompass_Preferred
A PAF receptor agonist with potency comparable to PAF C-18 and PAF C-16; chromogenic substrate used in Cayman’s PAF-AH Assay Kit (Item No. 760901)
Catalog No. 50-289-4449