Learn More
Cayman Chemical ANTIBODY 306-O12B-3 10mg
SDP
Supplier:  Cayman Chemical 3709610 MG
Encompass_Preferred
An ionizable cationic lipidoid; has been used in the generation of LNPs for the delivery of ASOs; intravenous administration of LNPs containing 306-O12B-3 localize to mouse liver; LNPs containing 306-O12B-3 in combination with other lipids encapsulating PCSK9-targeting ASOs decrease hepatic PCSK9 expression in mice; intravenous administration of LNPs containing 306-O12B-3 and NT1-O14B encapsulating ASOs against tau decreases tau expression and levels in mouse brain
Catalog No. 50-291-5238