Learn More
Cayman Chemical ANTIBODY 4E1RCat 10mg
SDP
Supplier:  Cayman Chemical 1839910 MG
Encompass_Preferred
An inhibitor of the eIF4F translation initiation complex that inhibits cap-dependent translation with an IC50 value of ~4 μM; reverses tumor chemoresistance in a mouse model of lymphoma by sensitizing cells to the proapoptotic action of DNA damage
Catalog No. 50-297-9658