Learn More
Cayman Chemical ANTIBODY 4E2RCat 5mg
SDP
Supplier:  Cayman Chemical 358945 MG
Encompass_Preferred
An inhibitor of the eIF4E-eIF4G protein-protein interaction (IC50 = 13.5 µM); reduces mRNA translation without inducing apoptosis in L-132 cells at 12.5 µM; decreases intracellular and extracellular SARS-COV titers and the percentage of cells expressing spike glycoprotein in SARS-COV-infected L-132 cells at 6 µM
Catalog No. 50-290-9242